4-(Methylthio)phenol 97%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO127130250EA
109
EUR
ACRO127130250
4-(Methylthio)phenol 97%
4-(Methylthio)phenol
Formel:
C₇H₈OS Molecular Weight: 140,21 g/mol Siedepunkt: 153…156 °C (20Torr) Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002351 CAS-Nummer: 1073-72-9 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Acidity | ≤0.2 % (as Sulfuric acid) |
Titration | 99.0 to 101.0 % (Hydrolysis) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...