Cooling Plate, CP 3523 (Klinipath)
Lieferant: Klinipath
KLINKP3523EA 2030 EUR
KLINKP3523
Cooling Plate, CP 3523 (Klinipath)
Robust and low noise cooling plate made of stainless steel and equipped with internal temperature controller with digital readout.
  • Integrates into laboratory bench thereby saving space
  • Large cooling capacity during continuous operation
  • Temperature settings from −15 to +15 °C
W×D×H: 400×645×250 mm
Bestellinformation: Optional control unit for the cooling plate can be ordered separately.