Adrenochrome 98%
|
Achtung
|
ACRO426212500EA
333.84
EUR
ACRO426212500
ACRO426210010
Adrenochrome 98%
Adrenochrome
Appearance: Dark red Crystalline solid
Formel:
C₉H₉NO₃ Lagertemperatur: Gefrierschrank |
CAS-Nummer:
54-06-8 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Dark red Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 115 to 120 °C |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...