2-Piperazinon 95%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO398710050EA
209
EUR
ACRO398710050
ACRO398710010
2-Piperazinon 95%
2-Piperazinon
Formel:
C₄H₈N₂O Molecular Weight: 100,12 Schmelzpunkt: 132…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318687 CAS-Nummer: 5625-67-2 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder or solid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...