Benchtop freezers, Revco®
Lieferant: Thermo Scientific
Revco®
REVCULT185-5-VEA 10120 EUR
REVCULT185-5-V
Benchtop freezers, Revco®
Gefriergeräte Ultratiefkühlgeräte
This benchtop freezez is ideal for personal or clinical trial applications.
  • Thermal interior sublid
  • Key operated main power switch
  • Steel cabinet with high epoxy finish
  • Solid state control system with adjustable temperature set point
  • Integrated with preset over temperature safety alarm with battery back-up and audible warning of temperature deviation
  • Also installed with air cooled refrigeration systems that are engineered to maintain design temperatures in warm 32 °C ambient conditions
Temperature range: −50 to −80 °C.