Syringes, tuberculin, Plastipak™
Lieferant: BD Medical
Plastipak™
613-5399EA 37.8 EUR
613-5399
Syringes, tuberculin, Plastipak™
Spritzen Spritzen, medizinisch
Tuberculin syringes without needles.
  • Syringe barrel and piston made of polypropylene
  • Single use
  • Sterile individual packaging
  • Low dose syringes for accurate measurements of small dose