Mass spectrometry sample prep kit, Pierce™
Lieferant: Thermo Fisher Scientific
PIER84840EA 636 EUR
PIER84840
Mass spectrometry sample prep kit, Pierce™
Pierce™ Mass Spec Sample Prep Kit for cultured cells is an easy-to-use, comprehensive kit for preparation of clean peptide mixtures from cultured cells for mass spectrometry (MS) analysis.
  • Reproducible results even for non-expert mass spectrometry analysis
  • Adapted to handle sample sizes between 10 to 200 µg
  • Percentage of missed cleavages is less than 10%
  • Cysteine reduction and alkylation are 100% with less than 1% over alkylation of non-cysteine residues
Lieferumfang: Kit contains proprietary lysis and digestion buffers, digestion indicator, DTT, iodoacetamide, Lys-C, and trypsin.