Bovine fibrinogen (from Plasma)

Lieferant: MP Biomedicals
ICNA0215112205EA 582 EUR
ICNA0215112205 ICNA0215112201
Bovine fibrinogen (from Plasma)
Proteine und Peptide
  • Protien content ~75%
  • Soluble in 5 to 7 minutes (4% aq soln)
  • Moisture content: <5%
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR