TES, 0,2 M buffer soln., pH 7,0

Lieferant: Thermo Fisher Scientific
J63379.AKEA 73.42 EUR
J63379.AK J63379.AE
TES, 0,2 M buffer soln., pH 7,0
Puffer Biologische Puffer
CAS No.: 70331-82-7
Physical Form: Liquid
MDL No.: MFCD00065482
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR