Succinate, 0,2 M buffer solution, pH 5,5

Lieferant: Thermo Fisher Scientific
J61100.APEA 66.46 EUR
J61100.AP J61100.AK
Succinate, 0,2 M buffer solution, pH 5,5
Puffer Biologische Puffer
CAS No.: 150-90-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002789
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR