Phosphate, 0,2 M buffer solution, pH 9,5

Lieferant: Thermo Fisher Scientific
J60819.K2EA 54.6 EUR
J60819.K2 J60819.AP
Phosphate, 0,2 M buffer solution, pH 9,5
Puffer Biologische Puffer
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR