Phosphate, 0,5M buffer solution, pH 7,4
Lieferant: Thermo Fisher Scientific
J60785.AKEA 81 EUR
J60785.AK J60785.AP
Phosphate, 0,5M buffer solution, pH 7,4
Puffer Biologische Puffer
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527