4-Methyl-1-naphthoyl chloride 98%

Gefahr

H64917.14EA 332.8 EUR
H64917.14
4-Methyl-1-naphthoyl chloride 98%
4-Methyl-1-naphthoyl chloride
Formel: C₁₂H₉ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD12025362
CAS-Nummer: 87700-67-2
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR