5-Bromo-2-(trifluoromethyl)pyrimidine 95%
|
Achtung
|
H64899.03EA
608.4
EUR
H64899.03
5-Bromo-2-(trifluoromethyl)pyrimidine 95%
5-Bromo-2-(trifluoromethyl)pyrimidine
Formel:
C₅H₂BrF₃N₂ Molecular Weight: 226,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09754046 CAS-Nummer: 799557-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...