3-Amino-6-brom-2-methylpyridin 98%

Achtung

H64756.14EA 347 EUR
H64756.14
3-Amino-6-brom-2-methylpyridin 98%
3-Amino-6-bromo-2-methylpyridine
Formel: C₆H₇BrN₂
Schmelzpunkt: 110…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095086
CAS-Nummer: 126325-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR