3-(Aminomethyl)benzoesäure Hydrochlorid 95%

Lieferant: Thermo Scientific
Achtung

H64546.03EA 85.38 EUR
H64546.03
3-(Aminomethyl)benzoesäure Hydrochlorid 95%
3-(Aminomethyl)benzoesäure Hydrochlorid
Formel: C₈H₁₀ClNO₂
Schmelzpunkt: 242…244 °C
MDL: MFCD03791117
CAS-Nummer: 876-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR