α,3-Dibrom-5-fluortoluol 95%
Lieferant: Thermo Scientific
|
Gefahr
|
H63769.06EA
166
EUR
H63769.06
α,3-Dibrom-5-fluortoluol 95%
α,3-Dibrom-5-fluortoluol
Formel:
C₇H₅Br₂F Molecular Weight: 267,92 g/mol |
MDL:
MFCD09258959 CAS-Nummer: 216755-57-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...