Z-D-Glu(OtBu)-OH 98%

Lieferant: Thermo Scientific

H63371.06EA 142 EUR
H63371.06
Z-D-Glu(OtBu)-OH 98%
Z-D-Glu(OtBu)-OH
Formel: C₁₇H₂₃NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00077018
CAS-Nummer: 51644-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR