L-Glutaminsäure-5-mthylester 95%
H63168.14EA
158.08
EUR
H63168.14
L-Glutaminsäure-5-mthylester 95%
L-Glutaminsäure-5-mthylester
Formel:
C₇H₁₃NO₄ Molecular Weight: 175,2 g/mol Schmelzpunkt: 179 °C (Zersetzung) |
MDL:
MFCD00037217 CAS-Nummer: 1119-33-1 EINECS: 214-274-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...