3-(Hydroxymethyl)-5-methylisoxazol 97%

Lieferant: Thermo Scientific
Achtung

H61948.MDEA 35.2 EUR
H61948.MD H61948.03
3-(Hydroxymethyl)-5-methylisoxazol 97%
3-(Hydroxymethyl)-5-methylisoxazole
Formel: C₅H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00085129
CAS-Nummer: 35166-33-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR