Ethyl-2-oxopyrrolidine-3-carboxylate 95%

Lieferant: Thermo Scientific
Warning

Synonyme: Ethyl 2-oxo-pyrrolidine-3-carboxylate

H57906.MD H57906.03
H57906.MDEA 51.1 EUR
H57906.MD H57906.03
Ethyl-2-oxopyrrolidine-3-carboxylate 95%
Ethyl-2-oxopyrrolidine-3-carboxylate
Formel: C₇H₁₁NO₃
Molecular Weight: 157,17 g/mol
Schmelzpunkt: 71…75 °C
MDL: MFCD06796534
CAS-Nummer: 36821-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR