3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole 95%
Lieferant: Thermo Scientific
|
Achtung
|
H32169.03EA
204
EUR
H32169.03
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole 95%
3-Chloro-4-(pyridin-3-yl)-1,2,5-thiadiazole
Formel:
C₇H₄ClN₃S Schmelzpunkt: 56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03066187 CAS-Nummer: 131986-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...