Barium metaborate monohydrate

Achtung

45823.30EA 16.1 EUR
45823.30
Barium metaborate monohydrate
Barium metaborate monohydrate
Formel: B₂BaO₄·H₂O
Molecular Weight: 240,97 g/mol (222,96 anhy)
Schmelzpunkt: >900 °C
Dichte: 3,35
Lagertemperatur: Raumtemperatur
MDL: MFCD00046182
CAS-Nummer: 19004-06-9
EINECS: 237-222-4
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR