Affinity chromatography columns, tagged package, GST
Lieferant: Cytiva
29-0588-04EA 122 EUR
29-0588-04
Affinity chromatography columns, tagged package, GST
Chromatographiesäulen
The GST package is designed for easy purification of GST-tagged proteins.
The GSTrap™ 4B column (1×1 ml) has a high binding capacity for GST-tagged proteins, giving you higher yields. A HiTrap™ desalting column (1×5 ml) is provided for rapid buffer exchange.
Lieferumfang: The kit contains one of each GSTrap™ 4B (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.