Anti-UFC1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BOSSBS-8372REA 691 EUR
BOSSBS-8372R
Anti-UFC1 Rabbit Polyclonal Antibody
Antikörper
E2-like enzyme which forms an intermediate with UFM1 via a thioester linkage.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UFC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR