Anti-Haspin Rabbit Polyclonal Antibody

Lieferant: Bioss
BOSSBS-15413REA 325 EUR
BOSSBS-15413R
Anti-Haspin Rabbit Polyclonal Antibody
Haspins is a protein family containing a distinctive C-terminal kinase domain and a divergent N-terminus. Haspin homologues occur within a diverse group of eukaryotes, including animals, plants and fungi, which suggests an early evolutionary origin. Haspin, a nuclear protein strongly expressed in male germ cells, is responsible for the phosphorylation of histone H3 at Thr-3. Depletion of Haspin RNA prevents normal alignment of chromosomes at metaphase, suggesting a crucial role for haspin during chromosome segregation. Expression of haspin also occurs in adult thymus and bone marrow, with weaker expression in adult prostate, intestine, lung, spleen, and lymph node. The gene encoding human haspin maps to chromosome 17p13.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Haspin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR