Anti-GPR37L1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BOSSBS-15390REA 691 EUR
BOSSBS-15390R
Anti-GPR37L1 Rabbit Polyclonal Antibody
Antikörper
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR37L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR