Anti-TIPIN Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB29474EA 518 EUR
ABNOPAB29474
Anti-TIPIN Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant human TIPIN.

Type: Primary
Antigen: TIPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR