Anti-CD86 Mouse Monoclonal Antibody [clone: 4H4]

Lieferant: Abnova
ABNOMAB5000-M02EA 344 EUR
ABNOMAB5000-M02
Anti-CD86 Mouse Monoclonal Antibody [clone: 4H4]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CD86.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR