Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 7H5]
Lieferant: Abnova
ABNOH00008600-M24EA 394 EUR
ABNOH00008600-M24
Anti-TNFSF11 Mouse Monoclonal Antibody [clone: 7H5]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TNFSF11.
Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 7H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human