Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 1G1]

Lieferant: Abnova
ABNOH00023495-M14EA 394 EUR
ABNOH00023495-M14
Anti-TNFRSF13B Mouse Monoclonal Antibody [clone: 1G1]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TNFRSF13B.

Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR