Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 8F3]

Lieferant: Abnova
ABNOMAB1297-M06EA 344 EUR
ABNOMAB1297-M06
Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 8F3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TNFRSF10A.

Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 8F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR