Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 4E10]
Lieferant: Abnova
ABNOH00027242-M50EA 394 EUR
ABNOH00027242-M50
Anti-TNFRSF21 Mouse Monoclonal Antibody [clone: 4E10]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TNFRSF21.
Type: Primary
Antigen: TNFRSF21
Clonality: Monoclonal
Clone: 4E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human