Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 3B1]

Lieferant: Abnova
ABNOH00009966-M03EA 394 EUR
ABNOH00009966-M03
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: 3B1]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TNFSF15.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR