Gas detector for CO
Lieferant: Testo
TEST0632.3155EA 411 EUR
TEST0632.3155
Gas detector for CO
This measuring instrument for recording ambient CO values identifies the slightest concentrations of the highly toxic gas. It allows the user to safely judge whether heating systems can be operated without danger.
  • Accuracy: ±3 ppm (0 to 20 ppm); ±5 ppm over 20 ppm
  • Optical and audible alarm when limit values are exceeded (freely adjustable limit values)
  • Printout of measurement data possible directly on site
An electrochemical CO sensor allows for reliable and highly precise measurement results.
Standards: TÜV-tested EN 50543