SPE manifolds, Resprep®
Lieferant: Restek
Resprep®
REST26077EA 0 EUR
REST26077 REST26080
SPE manifolds, Resprep®
Verteiler
Manifolds are used with any standard male Luer end SPE cartridge.
  • Inert PTFE sample guides reduce cross-contamination and carryover
  • Flexible sample collection rack will accommodate a variety of receiving vessels
  • Quick vacuum-release valve for better system control
  • Individual valves allow vacuum control for each cartridge
Lieferumfang: SPE manifolds are supplied with glass basin, built-in vacuum regulator, PP lid with individual control valves, collection rack and PTFE sample guides.