Protective hoods with visor for PAPR, full, FH2
Lieferant: Scott Safety
SCTS2022949EA 100.06 EUR
SCTS2022949
Protective hoods with visor for PAPR, full, FH2
Ultra light weight and comfortable full hood with integrated eye protection. This FH2 has been designed with a unique neoprene face seal to deliver the highest level of PAPR protection without compromising user comfort.
  • Revolutionary soft head cradle for intuitive and straightforward donning
  • Ultra clear PETG visor giving wide fi eld of view and a clear down view panel
  • Full hood design with skin-friendly neoprene chin seal
Standards: EN12941 TH3, EN166 1F and AS/ NZS1716-P3