Anti-HDAC5 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-J21EA 783 EUR
ROCK600-401-J21
Anti-HDAC5 Rabbit Polyclonal Antibody
Antikörper
Anti-HDAC5 antibody is useful for ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~124 kDa corresponding to the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000 to 1:60,000; Immunofluorescence: 1:100 to 1:500; Western Blot: 0,2 µg/ml; Immunochemistry: 1:100 to 1:500

Type: Primary
Antigen: HDAC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR