Anti-BCS1L Antibody Pair

Lieferant: Abnova
ABNOH00000617-AP51EA 835 EUR
ABNOH00000617-AP51
Anti-BCS1L Antibody Pair
Antikörper
This Set comes with matched Antibody Pair to detect and quantify protein level of human BCS1L.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-BCS1L (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-BCS1L (50 µg);

Type:
Antigen: BCS1L
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR