Anti-CASP3 + CDKN1A Antibody Pair

Lieferant: Abnova
ABNODI0085EA 668 EUR
ABNODI0085
Anti-CASP3 + CDKN1A Antibody Pair
Antikörper
This protein protein interaction Antibody Pair set comes with two antibodies to detect the protein-protein interaction, one against the CASP3 protein, and the other against the CDKN1A protein for use in Proximity Ligation Assay

Antibody pair set content: 1. CASP3 Rabbit purified Polyclonal Antibody (20 µg) 2. CDKN1A Mouse Monoclonal Antibody (40 µg) *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: CASP3 + CDKN1A
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR