Anti-RELA Antibody Pair

Lieferant: Abnova
ABNODP0217EA 835 EUR
ABNODP0217
Anti-RELA Antibody Pair
Antikörper
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the RELA protein, and the other against the specific S311 phosphorylated site of RELA for use in Proximity Ligation Assay

Antibody pair set content: 1. Phospho-RELA S311 Rabbit Polyclonal Antibody (20 µl) In PBS, 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. RELA Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.

Type:
Antigen: RELA
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR