Dehydrated Culture Media, Sifin, yeast
Lieferant: Sifin Diagnostics GmbH
715-9651EA 125 EUR
715-9651 715-9647 715-9646 715-9687 715-9592 715-9661 715-9649 SIFITN1047 SIFITN1264 715-9611 SIFITN1067 715-9577 715-9686
Dehydrated Culture Media, Sifin, yeast
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
These culture media and supplements are to cultivate the microorganisms or specifically to promote the growth of yeast and moulds. Thus, this is the optimal equippment for the analysis of veterinary or human samples.
  • Special media for food, water, pharmaceutical products
  • A vide range of different medias