Digital GelCam, EdvoFoto™
EDVO551EA 754 EUR
EDVO551
Digital GelCam, EdvoFoto™
An integrated system comprising an easy-to-use digital camera and a specially designed hood to provide a low cost alternative for gel photos. Will accommodate gels up to 7×14 cm. Photos may be downloaded to a computer.