Anti-Cnga1/Cnga3 Mouse Monoclonal Antibody (Biotin) [clone: S36-12]

Lieferant: Abnova
ABNOMAB11703EA 545 EUR
ABNOMAB11703
Anti-Cnga1/Cnga3 Mouse Monoclonal Antibody (Biotin) [clone: S36-12]
Antikörper
Mouse monoclonal antibody raised against partial synthetic protein of Cng.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: Cnga1/Cnga3
Clonality: Monoclonal
Clone: S36-12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR