Anti-DUX4 Mouse Monoclonal Antibody (ATTO 594) [clone: P2B1]
Lieferant: Abnova
ABNOMAB11446EA 772 EUR
ABNOMAB11446
Anti-DUX4 Mouse Monoclonal Antibody (ATTO 594) [clone: P2B1]
Antikörper
Mouse monoclonal antibody raised against recombinant partial DUX4.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DUX4
Clonality: Monoclonal
Clone: P2B1
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human