Anti-MDC1 Mouse Monoclonal Antibody [clone: P2B11]
Lieferant: Abnova
ABNOMAB11460EA 518 EUR
ABNOMAB11460
Anti-MDC1 Mouse Monoclonal Antibody [clone: P2B11]
Antikörper
Mouse monoclonal antibody raised against recombinant partial Mdc1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: MDC1
Clonality: Monoclonal
Clone: P2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chimpanzee, Human, Mouse