Anti-CARS Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28664EA 518 EUR
ABNOPAB28664
Anti-CARS Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant CARS.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:200; Immunofluorescence: 1-4 ?g/ml; Western Blot: 1:100-1:250

Type: Primary
Antigen: CARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR