Anti-PSMC4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28661EA 568 EUR
ABNOPAB28661
Anti-PSMC4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant PSMC4.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:200-1:500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: PSMC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR