Anti-AADAC Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28608EA 568 EUR
ABNOPAB28608
Anti-AADAC Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant AADAC.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:100-1:250

Type: Primary
Antigen: AADAC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR