Anti-TENM1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28599EA 568 EUR
ABNOPAB28599
Anti-TENM1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant ODZ1.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20

Type: Primary
Antigen: TENM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR